ENAH11DD1F94 Enamine

2,2-difluoro-2-(4-hydroxypiperidin-4-yl)acetamide hydrochloride

  • MDL number MFCD31666129
  • Cas Number : 2229161-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721720
Availability details for
Quantity
  • Availability
  • Add to Cart