ENAH11DD1F95 Enamine

methyl 2-amino-5-bromo-6-(trifluoromethyl)pyridine-3-carboxylate

  • MDL number MFCD31665388
  • Cas Number : 2225146-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720969
Availability details for
Quantity
  • Availability
  • Add to Cart