ENAH11DD1F9D Enamine

4,4-difluoro-3-(2-fluorophenyl)butanoic acid

  • MDL number MFCD31665389
  • Cas Number : 1897774-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1720987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720987
Availability details for
Quantity
  • Availability
  • Add to Cart