ENAH11DD1FC8 Enamine

7-fluoro-1,2,3,4-tetrahydroquinolin-4-ol

  • MDL number MFCD28111854
  • Cas Number : 1824060-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721917
Availability details for
Quantity
  • Availability
  • Add to Cart