ENAH11DD1FD4 Enamine

1-(2-amino-1,3-thiazol-5-yl)ethan-1-one hydrobromide

  • MDL number MFCD29921987
  • Cas Number : 88158-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Product Link https://www.enaminestore.com/catalog/EN300-1721939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721939
Availability details for
Quantity
  • Availability
  • Add to Cart