ENAH11DD1FD5 Enamine

1,2,3,4-tetrahydroquinoline-1-carbonyl chloride

  • MDL number MFCD19281935
  • Cas Number : 2616-50-4
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,4-dihydro-1(2H)-quinolinecarbonyl chloride
Inchi Code 1S/C10H10ClNO/c11-10(13)12-7-3-5-8-4-1-2-6-9(8)12/h1-2,4,6H,3,5,7H2
Inchi Key KRGMMKVSHXVQCM-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1721959

Safety Information

signalword Danger
pcodes P234,P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P390,P403+P233,P405,P406,P501
MSDS https://www.enaminestore.com/catalog/EN300-1721959
pictograms GHS05,GHS07
hstatements H290,H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart