ENAH11DD1FDE Enamine

2-cyclopropyl-4,6-dimethylpyrimidine-5-carboxylic acid

  • MDL number MFCD16868506
  • Cas Number : 548773-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1721033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721033
Availability details for
Quantity
  • Availability
  • Add to Cart