ENAH11DD1FDF Enamine

6-hydroxy-8-methyl-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD24616311
  • Cas Number : 1239768-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1721981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721981
Availability details for
Quantity
  • Availability
  • Add to Cart