ENAH11DD1FE0 Enamine

2-bromo-3-isothiocyanatopyridine

  • MDL number MFCD22557542
  • Cas Number : 1360887-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721984
Availability details for
Quantity
  • Availability
  • Add to Cart