ENAH11DD1FE4 Enamine

1-methyl-3-(1-methyl-1H-pyrazol-5-yl)-1H-pyrazol-4-amine dihydrochloride

  • MDL number MFCD31666820
  • Cas Number : 2228903-62-4
  • Molecular Weight : 250.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,2'-dimethyl-1H,2'H-[3,3'-bipyrazol]-4-amine dihydrochloride
Inchi Code 1S/C8H11N5.2ClH/c1-12-5-6(9)8(11-12)7-3-4-10-13(7)2;;/h3-5H,9H2,1-2H3;2*1H
Inchi Key OSKJWZXFPISUFJ-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1721990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721990
Availability details for
Quantity
  • Availability
  • Add to Cart