ENAH11DD1FE9 Enamine

4-bromo-1-tert-butyl-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD16092749
  • Cas Number : 1251118-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1722006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722006
Availability details for
Quantity
  • Availability
  • Add to Cart