ENAH11DD1FEA Enamine

4-[(1S)-1-hydroxyethyl]benzonitrile

  • MDL number MFCD08702986
  • Cas Number : 101219-71-0
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[(1S)-1-hydroxyethyl]benzonitrile
Inchi Code 1S/C9H9NO/c1-7(11)9-4-2-8(6-10)3-5-9/h2-5,7,11H,1H3/t7-/m0/s1
Inchi Key XGAVOODMMBMCKV-ZETCQYMHSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1722008

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1722008
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart