ENAH11DD1FEC Enamine

5-(2-bromoethynyl)-3-methoxy-1,2-thiazole

  • MDL number MFCD31666784
  • Cas Number : 2228572-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1722013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722013
Availability details for
Quantity
  • Availability
  • Add to Cart