ENAH11DD1FEF Enamine

2-chloro-4-(methylsulfanyl)benzoic acid

  • MDL number MFCD17256624
  • Cas Number : 1079360-49-8
  • Molecular Weight : 202.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-4-(methylsulfanyl)benzoic acid
Inchi Code 1S/C8H7ClO2S/c1-12-5-2-3-6(8(10)11)7(9)4-5/h2-4H,1H3,(H,10,11)
Inchi Key GRUJNPZAMCELAI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1722018

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1722018
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart