ENAH11DD1FF1 Enamine

methyl bicyclo[3.1.0]hexane-3-carboxylate

  • MDL number MFCD23907759
  • Cas Number : 90199-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1722022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722022
Availability details for
Quantity
  • Availability
  • Add to Cart