ENAH11DD1FF7 Enamine

(2S)-2-(4-bromophenyl)-4-methylmorpholine

  • MDL number MFCD27932933
  • Cas Number : 920798-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1721067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721067
Availability details for
Quantity
  • Availability
  • Add to Cart