ENAH11DD1FF8 Enamine

ethyl 3-(4-fluorophenyl)-2-methylbutanoate

  • MDL number MFCD31666861
  • Cas Number : 2107198-78-5
  • Molecular Weight : 224.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-(4-fluorophenyl)-2-methylbutanoate
Inchi Code 1S/C13H17FO2/c1-4-16-13(15)10(3)9(2)11-5-7-12(14)8-6-11/h5-10H,4H2,1-3H3
Inchi Key ADESQKOOUKEVNB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1722031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722031
Availability details for
Quantity
  • Availability
  • Add to Cart