ENAH11DD1FFC Enamine

decane-1,9-diol

  • MDL number MFCD31665384

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30-33
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1720826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720826
Availability details for
Quantity
  • Availability
  • Add to Cart