ENAH11DD1FFE Enamine

9-methyl-9-azabicyclo[4.2.1]nonan-3-one

  • Molecular Weight : 153.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 9-methyl-9-azabicyclo[4.2.1]nonan-3-one
Inchi Code 1S/C9H15NO/c1-10-7-2-3-8(10)6-9(11)5-4-7/h7-8H,2-6H2,1H3
Inchi Key IBOXGLLKSPCYLZ-UHFFFAOYSA-N

Description

Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1722039

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1722039
pictograms GHS05,GHS07
hstatements H302,H312,H315,H318,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart