ENAH11DD200C Enamine

6-amino-2-(oxan-4-yl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD21296007
  • Cas Number : 1488002-44-3
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-2-tetrahydro-2H-pyran-4-yl-4(3H)-pyrimidinone
Inchi Code 1S/C9H13N3O2/c10-7-5-8(13)12-9(11-7)6-1-3-14-4-2-6/h5,11H,1-4,10H2,(H,12,13)
Inchi Key CLRVNOVKOWPRAD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1721116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721116
Availability details for
Quantity
  • Availability
  • Add to Cart