ENAH11DD200D Enamine

4,4-difluoropiperidine-3-carbonitrile hydrochloride

  • MDL number MFCD31666814
  • Cas Number : 2116938-91-9
  • Molecular Weight : 182.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,4-difluoropiperidine-3-carbonitrile hydrochloride
Inchi Code 1S/C6H8F2N2.ClH/c7-6(8)1-2-10-4-5(6)3-9;/h5,10H,1-2,4H2;1H
Inchi Key GUEBRQPAXGKBQM-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1722110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722110
Availability details for
Quantity
  • Availability
  • Add to Cart