ENAH11DD200F Enamine

4-[3-(trifluoromethyl)-1H-pyrazol-5-yl]aniline

  • MDL number MFCD31666815
  • Cas Number : 2171036-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1722114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722114
Availability details for
Quantity
  • Availability
  • Add to Cart