ENAH11DD2012 Enamine

4-methyl-2-(pyrrolidin-2-yl)-1,3-oxazole hydrochloride

  • MDL number MFCD31666846
  • Cas Number : 2229325-84-0
  • Molecular Weight : 188.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methyl-2-(pyrrolidin-2-yl)oxazole hydrochloride
Inchi Code 1S/C8H12N2O.ClH/c1-6-5-11-8(10-6)7-3-2-4-9-7;/h5,7,9H,2-4H2,1H3;1H
Inchi Key OXNOCQMAGMGKFX-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1722122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722122
Availability details for
Quantity
  • Availability
  • Add to Cart