ENAH11DD2035 Enamine

3-amino-2,5-dihydrofuran-2-one

  • MDL number MFCD15145812
  • Cas Number : 33693-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722218
Availability details for
Quantity
  • Availability
  • Add to Cart