ENAH11DD2055 Enamine

2-amino-4-bromo-6-(trifluoromethyl)phenol

  • MDL number MFCD28741409
  • Cas Number : 1805552-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722344
Availability details for
Quantity
  • Availability
  • Add to Cart