ENAH11DD2056 Enamine

6-bromo-1,2,3-benzothiadiazole

  • MDL number MFCD29110700
  • Cas Number : 31860-02-3
  • Molecular Weight : 215.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 67-70
Iupac Name 6-bromobenzo[d][1,2,3]thiadiazole
Inchi Code 1S/C6H3BrN2S/c7-4-1-2-5-6(3-4)10-9-8-5/h1-3H
Inchi Key HRDFWZRRQDWWAB-UHFFFAOYSA-N

Description

Melting Point 67-70
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1721144

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1721144
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart