ENAH11DD2057 Enamine

(4-cyano-3-cyclopropylphenyl)boronic acid

  • MDL number MFCD21099622
  • Cas Number : 2225155-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1721147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721147
Availability details for
Quantity
  • Availability
  • Add to Cart