ENAH11DD205C Enamine

ethyl 3-amino-3-(1,3-thiazol-2-yl)propanoate dihydrochloride

  • MDL number MFCD31666295
  • Cas Number : 2229402-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-1722369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722369
Availability details for
Quantity
  • Availability
  • Add to Cart