ENAH11DD2064 Enamine

4-oxothiolane-2-carboxylic acid

  • MDL number MFCD18817140
  • Cas Number : 89323-44-4
  • Molecular Weight : 146.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 134-138
Iupac Name 4-oxotetrahydrothiophene-2-carboxylic acid
Inchi Code 1S/C5H6O3S/c6-3-1-4(5(7)8)9-2-3/h4H,1-2H2,(H,7,8)
Inchi Key IBSKPYFOLKUHNM-UHFFFAOYSA-N

Description

Melting Point 134-138
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1722378

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1722378
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart