ENAH11DD2065 Enamine

(1R)-1-(4-bromophenyl)-2,2-difluoroethan-1-amine hydrochloride

  • MDL number MFCD26569115
  • Cas Number : 2225127-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-1721153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721153
Availability details for
Quantity
  • Availability
  • Add to Cart