ENAH11DD2067 Enamine

(2-chloro-3-fluoropyridin-4-yl)methanamine hydrochloride

  • MDL number MFCD31665456
  • Cas Number : 2225146-88-1
  • Molecular Weight : 197.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chloro-3-fluoropyridin-4-yl)methanamine hydrochloride
Inchi Code 1S/C6H6ClFN2.ClH/c7-6-5(8)4(3-9)1-2-10-6;/h1-2H,3,9H2;1H
Inchi Key DUWBCOMVESTEAF-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1721154

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1721154
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart