ENAH11DD2068 Enamine

3-fluorooxane-3-carboxylic acid

  • MDL number MFCD28635743
  • Cas Number : 1547757-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1722390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722390
Availability details for
Quantity
  • Availability
  • Add to Cart