ENAH11DD2069 Enamine

N2-methyl-N2-[(1-methyl-1H-imidazol-2-yl)methyl]pyrimidine-2,5-diamine

  • MDL number MFCD17275441
  • Cas Number : 1339627-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722392
Availability details for
Quantity
  • Availability
  • Add to Cart