ENAH11DD206C Enamine

3-(dimethylamino)-4-(methylamino)cyclobut-3-ene-1,2-dione

  • MDL number MFCD20680442
  • Cas Number : 162362-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1720841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1720841
Availability details for
Quantity
  • Availability
  • Add to Cart