ENAH11DD2070 Enamine

(2S,3R)-2-amino-3-hydroxybutanedioic acid hydrochloride

  • MDL number MFCD31665457
  • Cas Number : 1848942-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-1721165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721165
Availability details for
Quantity
  • Availability
  • Add to Cart