ENAH11DD2072 Enamine

methyl 2-amino-3-(ethylsulfanyl)propanoate

  • MDL number MFCD16143962

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1722416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722416
Availability details for
Quantity
  • Availability
  • Add to Cart