ENAH11DD2073 Enamine

4-chloro-2-[3-(trifluoromethyl)phenyl]phenol

  • MDL number MFCD18423949
  • Cas Number : 178374-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1722417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722417
Availability details for
Quantity
  • Availability
  • Add to Cart