ENAH11DD2076 Enamine

5-bromo-2-(propan-2-yl)-1,3-oxazole

  • MDL number MFCD22682026
  • Cas Number : 1391739-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1722420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722420
Availability details for
Quantity
  • Availability
  • Add to Cart