ENAH11DD2077 Enamine

1-(2-hydroxy-4-methylphenyl)pentan-1-one

  • MDL number MFCD00127711
  • Cas Number : 173851-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1721167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721167
Availability details for
Quantity
  • Availability
  • Add to Cart