ENAH11DD207D Enamine

4-amino-5-methylthiophene-3-carboxylic acid hydrochloride

  • MDL number MFCD31666299
  • Cas Number : 2229531-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-1722461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722461
Availability details for
Quantity
  • Availability
  • Add to Cart