ENAH11DD2084 Enamine

[3-(tert-butoxy)-2,2-dimethylcyclobutyl]methanamine hydrochloride

  • MDL number MFCD31666300
  • Cas Number : 2228825-11-2
  • Molecular Weight : 221.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-(tert-butoxy)-2,2-dimethylcyclobutyl)methanamine hydrochloride
Inchi Code 1S/C11H23NO.ClH/c1-10(2,3)13-9-6-8(7-12)11(9,4)5;/h8-9H,6-7,12H2,1-5H3;1H
Inchi Key YMIQQOIOISKGBP-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1722469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722469
Availability details for
Quantity
  • Availability
  • Add to Cart