ENAH11DD2085 Enamine

5,5-dimethoxypiperidine-3-carboxamide hydrochloride

  • MDL number MFCD31666769
  • Cas Number : 2228908-29-8
  • Molecular Weight : 224.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5,5-dimethoxypiperidine-3-carboxamide hydrochloride
Inchi Code 1S/C8H16N2O3.ClH/c1-12-8(13-2)3-6(7(9)11)4-10-5-8;/h6,10H,3-5H2,1-2H3,(H2,9,11);1H
Inchi Key ZPWCLBBWFQLKKM-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1722470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722470
Availability details for
Quantity
  • Availability
  • Add to Cart