ENAH11DD208D Enamine

(1-cyclopropylethyl)(ethyl)amine hydrochloride

  • MDL number MFCD31666376
  • Cas Number : 2228829-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-1722479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722479
Availability details for
Quantity
  • Availability
  • Add to Cart