ENAH11DD2092 Enamine

6-(oxolan-2-yl)-4-(trifluoromethyl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD31666377
  • Cas Number : 2228135-30-4
  • Molecular Weight : 234.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(dihydrofuran-2(3H)-ylidene)-4-(trifluoromethyl)-1,6-dihydropyridazin-3(2H)-one
Inchi Code 1S/C9H9F3N2O2/c10-9(11,12)5-4-6(13-14-8(5)15)7-2-1-3-16-7/h4,13H,1-3H2,(H,14,15)
Inchi Key CWETZYBFSZUTBE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1722496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722496
Availability details for
Quantity
  • Availability
  • Add to Cart