ENAH11DD2093 Enamine

5-ethoxy-3-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD03011727
  • Cas Number : 57291-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-184
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1722504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722504
Availability details for
Quantity
  • Availability
  • Add to Cart