ENAH11DD2094 Enamine

5-ethyl-3-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD21985801
  • Cas Number : 1368639-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1722506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722506
Availability details for
Quantity
  • Availability
  • Add to Cart