ENAH11DD2096 Enamine

methyl 6-bromo-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD20265058
  • Cas Number : 1934399-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1722527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722527
Availability details for
Quantity
  • Availability
  • Add to Cart