ENAH11DD2097 Enamine

1-(2-fluoro-3-methylphenyl)-2-(pyridin-4-yl)ethan-1-one

  • MDL number MFCD25126310
  • Cas Number : 1504551-75-0
  • Molecular Weight : 229.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-fluoro-3-methylphenyl)-2-(pyridin-4-yl)ethan-1-one
Inchi Code 1S/C14H12FNO/c1-10-3-2-4-12(14(10)15)13(17)9-11-5-7-16-8-6-11/h2-8H,9H2,1H3
Inchi Key PZUVEIPVEVOPNP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1721200

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1721200
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart