ENAH11DD20E9 Enamine

1-(1H-1,3-benzodiazol-2-yl)azetidin-3-amine dihydrochloride

  • MDL number MFCD31666629
  • Cas Number : 2228650-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722780

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722780
Availability details for
Quantity
  • Availability
  • Add to Cart