ENAH11DD20FD Enamine

2-amino-2-(2,3-dihydro-1H-inden-1-yl)acetic acid hydrochloride

  • MDL number MFCD31666383
  • Cas Number : 2228094-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722849
Availability details for
Quantity
  • Availability
  • Add to Cart